Cell-Sorb Plus Animal Bedding
82040 82010 82020
470227-550BX 30.25 USD
470227-550 470227-546 470227-548
Cell-Sorb Plus Animal Bedding
Cell Sorb Plus is produced with recycled newspapers and a proprietary odor neutralizing mineral that is superior to zeolite and baking soda.
  • Available in multiple sizes
  • Environmentally friendly
  • Highly absorbent, cages remain dry
  • Up to 4 times as absorbent as wood shavings
Soft, granular paper bedding for animal cages is highly absorbent. Recommended for mice, rats, rabbits, and other small animals for use as contact and noncontact bedding. Recycled paper is processed, with heat and compression to destroy any hazardous organisms, it also 100% biodegradable and compostable. Provides for a clean and dry environment for your pet while completely eliminating ammonia odors for up to 3 weeks. Provides for a clean and dry environment for your pet while completely eliminating ammonia odors for up to 3 weeks. The pellets absorb urine and neutralize odor.
Directions: Place 1” to 2” of Cell Sorb Plus in cage. Stir litter every other day for longer life. Periodically remove soiled litter. Cell Sorb Plus will keep your pet dry and comfortable for up to 3 weeks without a complete litter.