Preserved Ambystoma
136.1050.10 136.1051.10
470227-582PK 99.99 USD
470227-582 470227-580
Preserved Ambystoma
Specimens Preserved Specimens Preserved Animal Materials
Formalin Preserved Ambystoma (Tiger Salamander)
  • Amphibian dissection
  • Developemental biology
  • Alternative to Necturus
Our Ambystoma (Tiger Salamanders) are great for studying developmental biology. Unlike their cousins the mudpuppy (Necturus), these salamanders do not exhibit neoteny and lose their juvenile characteristics as they mature. Supplied in Vack Packs or Pails of 10.
Similar to Necturs dissection. Great alternative.