Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Uvex Genesis® S Safety Eyewear, Honeywell Safety

Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
uvex genesis®
S3702 S3701 S3704 S3703 S3702X S3700D S3701X S3700X S3700
Uvex Genesis offers a 6% smaller frame for workers with slimmer facial profiles

  • Wrap-around lens for uncompromised peripheral vision
  • Padded temples and tips provide all-day comfort
  • Ultra-dura, scratch- and chemical-resistant hard coat is permanently bonded to the lens
  • Offer 99.9% ultra-violet protection
  • Flexible nasal fingers and ratcheting temples for customized fit
  • Economical and easy lens replacement system allows for multiple lens tints and extends the life of the eyewear

Ideal for use in construction, food services and pharmaceutical industries.

The soft browguard absorbs and diffuses impact, flexible fingers on nosepiece minimize slippage.

Certifications: Meets ANSI Z87.1-2010 and is certified to the requirements of the CSA Z94.3 standard.



 

ORDER

Expand All / Collapse All
Frame Material Description Frame Color Lens Coating Lens Tint Lens Material Supplier No. VWR Catalog Number Unit Price Quantity
Polycarbonate with elastomer Uvex Genesis S (Slim) Pewter/Black Ultra-dura Anti-scratch Clear Polycarbonate S3700 89095-368 Each Retrieving
Pack of 10 Retrieving
Polycarbonate with elastomer Uvex Genesis S (Slim) Pewter/Black Dura-streme Dual Anti-fog/Anti-Scratch Clear Polycarbonate S3700D 89095-372 Each Retrieving
Pack of 10 Retrieving
Polycarbonate with elastomer Uvex Genesis S (Slim) Pewter/Black Ultra-dura Anti-scratch Gray Polycarbonate S3701 89095-374 Each Retrieving
Pack of 10 Retrieving
Polycarbonate with elastomer Uvex Genesis S (Slim) Pewter/Black Ultra-dura Anti-scratch Amber Polycarbonate S3702 89095-378 Each Retrieving
Pack of 10 Retrieving
Polycarbonate with elastomer Uvex Genesis S (Slim) Pewter/Black Ultra-dura Anti-scratch SCT-Reflect 50 Polycarbonate S3703 89095-382 Each Retrieving
Pack of 10 Retrieving
Polycarbonate with elastomer Uvex Genesis S (Slim) Pewter/Black Ultra-dura Anti-scratch Dark Gray Polycarbonate S3704 89095-384 Each Retrieving
Pack of 10 Retrieving
Polycarbonate with elastomer Uvex Genesis S (Slim) Pewter/Black UV Extreme Anti-fog Clear Polycarbonate S3700X 89095-370 Each Retrieving
Pack of 10 Retrieving
Polycarbonate with elastomer Uvex Genesis S (Slim) Pewter/Black UV Extreme Anti-fog Gray Polycarbonate S3701X 89095-376 Each Retrieving
Pack of 10 Retrieving
Polycarbonate with elastomer Uvex Genesis S (Slim) Pewter/Black UV Extreme Anti-fog Amber Polycarbonate S3702X 89095-380 Each Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty