Copper (II) ethylenediamine 1 M

Supplier: GFS CHEMICALS INC

Synonyms: CupriethylendiamineBis(ethylenediamine)copper dihydroxideCopper(II)-ethylenediame complex

32101
34126-300EA 284.98 USD
34126-300
Copper (II) ethylenediamine 1 M
Copper (II) ethylenediamine

For determination of solution viscosity of pulps.

Formula: Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MW: 217.76 g/mol
MDL Number: MFCD00274628
CAS Number: 14552-35-3
UN: 1761
ADR: 8,II

Order Now

Specification Test Results

Specific Gravity 1.100
Ethylenediamine to copper ratio 2.00 +/- 0.04
Copper molarity 1.00 +/- 0.02 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR