You Searched For: Methoxycarbonyl glycine
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Ambeed
Description: 2-((Methoxycarbonyl)amino)acetic acid, Purity: 97%, CAS Number: 1670-97-9, Appearance: White to off-white solid, Storage: Sealed in dry, Room Temperature, Size: 5g