n-Tritriacontane ≥96%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-tritriacontane

05330025MG
100282-184EA 389.69 USD
100282-184
n-Tritriacontane ≥96%
n-Tritriacontane

Store in a cool, dry place

Formula: H₃C(CH₂)₃₁CH₃
MW: 464.9 g/mol
MDL Number: MFCD00010187
CAS Number: 630-05-7
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR