Tricosanoic acid

Supplier: INDOFINE CHEMICAL COMPANY MS

102300100MG
100299-066EA 329.54 USD
100299-066
Tricosanoic acid
Tricosanoic acid

Store in a cool, dry place

Formula: C₂₃H₄₆O₂
MW: 354.62 g/mol
MDL Number: MFCD00002808
CAS Number: 2433-96-7
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR