n-Pentadecane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-PentadecanePentadecane

051500100MG
100293-050EA 115.08 USD
100293-050
n-Pentadecane ≥99%
n-Pentadecane

Store in a cool, dry place

Formula: H₃C(CH₂)₁₃CH₃
MW: 212.42 g/mol
MDL Number: MFCD00008990
CAS Number: 629-62-9
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR