N-Octadecanoylsphingosylphosphorylcholine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

5610845MG
100291-338EA 1108.92 USD
100291-338
N-Octadecanoylsphingosylphosphorylcholine ≥98%
N-Octadecanoylsphingosylphosphorylcholine

Store in a cool, dry place

MW: 749 g/mol (C18:0)
MDL Number: MFCD02259130
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR