Methyl-3-hydroxyundecanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

24110350MG
100293-676EA 630.31 USD
100293-676
Methyl-3-hydroxyundecanoate ≥98%
Methyl-3-hydroxyundecanoate

Store in a cool, dry place

Formula: C₁₂H₂₄O₃
MDL Number: MFCD00673454
CAS Number: 129758-71-0
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR