1-Heptadecanol ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Heptadecan-1-olHexadecyl carbinol

401700100MG
100298-772EA 188.31 USD
100298-772
1-Heptadecanol ≥99%
1-Heptadecanol

Store in a cool, dry place

Formula: H₃C(CH₂)₁₅CH₂OH
MW: 256.47 g/mol
MDL Number: MFCD00002822
CAS Number: 1454-85-9
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR