D-erythro-Dihydrosphingosine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

5613075MG
100291-432EA 529.62 USD
100291-432
D-erythro-Dihydrosphingosine ≥98%
D-erythro-Dihydrosphingosine

Store in a cool, dry place

Formula: C₁₈H₃₉NO₂
MDL Number: MFCD07371371
CAS Number: 764-22-7
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR