Ceramide trihexosides (Cth,Gb3) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

5610551MG
100291-278EA 1503.85 USD
100291-278
Ceramide trihexosides (Cth,Gb3) ≥99%
Ceramide trihexosides (Cth,Gb3)

Store in a cool, dry place

MW: 1052 g/mol (C18:0)
MDL Number: MFCD00166520
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR