4-(5-Mercapto-4H-1,2,4-triazol-3-yl)phenol ≥98%, crystalline solid

Supplier: INDOFINE CHEMICAL COMPANY MS

250211GM
100269-532EA 47.08 USD
100269-532
4-(5-Mercapto-4H-1,2,4-triazol-3-yl)phenol ≥98%, crystalline solid
4-(5-Mercapto-4H-1,2,4-triazol-3-yl)phenol

Off white crystalline solid


Store in a cool, dry place

Formula: C₈H₇N₃OS
Melting Pt: 301…303 °C
MDL Number: MFCD01930057
CAS Number: 3414-96-8
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR