6-Methylchromone ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 6-Methyl-4H-chromen-4-one

191471GM
100264-016EA 31.38 USD
100264-016
6-Methylchromone ≥98%, powder
6-Methylchromone

Pale white powder


Store in a cool, dry place

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Melting Pt: 88…89 °C
MDL Number: MFCD00218598
CAS Number: 38445-23-7
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR