1-Palmitin-3-stearin ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-Palmitoyl-3-stearoyl-rac-glycerol

32165425MG
100290-322EA 657.77 USD
100290-322
1-Palmitin-3-stearin ≥98%
1-Palmitin-3-stearin

Store in a cool, dry place

Formula: C₃₇H₇₂O₅
MW: 596.90 g/mol
MDL Number: MFCD00079234
CAS Number: 17708-08-6
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR