INTRALIPID IV Fat Emulsion, BAXTER IV SOLUTIONS
Supplier: Baxter
2B6063 2B6064 2B6061 2B6062 2B6065
75849-312CS 585.41 USD
75849-312 75849-310 75849-316 75849-314 75849-308
INTRALIPID IV Fat Emulsion, BAXTER IV SOLUTIONS
Media for Microbiology Additives and Supplements for Microbiology
Intralipid® 20% Pharmacy Bulk Package is not intended for direct infusion
  • Does not contain PVC, DEHP or Plasticizer
It is a sterile dosage form which contains several single doses for use in the preparation of three‑in‑one or total nutrient admixtures (TNAs) in a pharmacy admixture program
It is made up of 20% Soybean Oil, 1.2% Egg Yolk Phospholipids, 2.25% Glycerin, and Water for Injection
Packaging: BIOFINE Container, Pharmacy Bulk Package
Caution: Please refer to full Prescribing Information, including Boxed Warning.