FluorLast™ Mounting Media, Biovision

Supplier: BioVision
FluorLast™
1218-20 1214-20 1213-20 1217-20
10003-038EA 136.26 USD
10003-038 10003-040 10003-042 10003-044
FluorLast™ Mounting Media, Biovision
Stains Microscopy and staining reagents
Mounting medium are prepared using a unique, stable formula to preserving fluorescence of various fluorescein tissues and cell smears.

  • Ready-to-use mounting medium
  • Applications: Immunofluorescence, confocal microscopy
  • Storage Temperature: 4° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR