ADIPSYN-1082
SYN-1082-M005 SYN-1082-M001 SYN-1082-M010 SYN-1082-M050
102991-902EA 96.92 USD
102991-902 102991-904 102991-900 102991-906
ADIPSYN-1082
Sorafenib is a multi-kinase inhibitor targeting a number of serine/threonine and receptor tyrosine kinases. It inhibits Raf-1, B-Raf and VEGFR-2 with IC(50) values of 6nM, 22nM and 90nM, respectively.