VTX-27 ≥95%
Supplier: ADIPOGEN CORP MS
SYN-1206-M005
SYN-1206-M010
SYN-1206-M001
102992-792EA
329.54
USD
102992-792
102992-796
102992-794
VTX-27 ≥95%
VTX-27
VTX-27 is a potent and selective inhibitor of the PKCtheta enzyme, with over 1000-fold against classical isoforms and over 10000-fold against atypical isoforms.
Formula:
C₂₀H₂₄ClFN₆O MW: 418.9 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD31618412 CAS Number: 1321924-70-2 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...