KW-2449 hydrochloride ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
[4-[2-(1H-Indazol-3-yl)ethenyl]phenyl]-1-piperazinylmethanone
SYN-1205-M001
SYN-1205-M050
SYN-1205-M010
SYN-1205-M005
102992-788EA
134.31
USD
102992-788
102992-790
102992-786
102992-784
KW-2449 hydrochloride ≥95%
KW-2449 hydrochloride
KW-2449 is a multiple-targeted inhibitor, mostly for Flt3 with IC(50) of 6.6nM, modestly potent to FGFR1, Bcr-Abl and Aurora A; little effect on PDGFRbeta, IGF-1R, EGFR.
Formula:
C₂₀H₂₀N₄O·HCl MW: 368.86 g/mol Storage Temperature: Freezer |
CAS Number:
841259-17-4 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...