Bafetinib ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
NS-187
SYN-1179-M010
SYN-1179-M001
SYN-1179-M005
SYN-1179-M050
102992-606EA
254.77
USD
102992-606
102992-604
102992-608
102992-610
Bafetinib ≥95%
Bafetinib
Bafetinib (INNO-406) is a potent and selective dual Bcr-Abl, Lyn inhibitor with IC(50) values of 5.8nM and 19nM, respectively.
Formula:
C₃₀H₃₁F₃N₈O MW: 576.6 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD18633200 CAS Number: 859212-16-1 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...