CEP-33779 ≥95%
Supplier: ADIPOGEN CORP MS
SYN-1156-M050
SYN-1156-M005
SYN-1156-M010
SYN-1156-M001
102992-426EA
217.38
USD
102992-426
102992-428
102992-430
102992-432
CEP-33779 ≥95%
CEP-33779
CEP-33779 is a highly selective, orally active inhibitor for JAK2 with an IC(50) of 1.8nM in enzyme and 61nM in cell activity. It inhibits JAK3 with an IC(50) of 150nM.
Formula:
C₂₄H₂₆N₆O₂S MW: 462.6 g/mol Storage Temperature: Freezer |
CAS Number:
1346168-57-7 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...