BMS-599626 ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
AC-480
SYN-1142-M001
SYN-1142-M010
SYN-1142-M005
SYN-1142-M050
102992-328EA
156.46
USD
102992-328
102992-330
102992-334
102992-332
BMS-599626 ≥95%
BMS-599626
BMS-599626 is an orally bioavailable, highly selective and potent pan-HER kinase inhibitor. It inhibits HER1 and HER2 with IC(50) values of 20 and 30nM, respectively.
Formula:
C₂₇H₂₇FN₈O₃ MW: 530.6 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD10565678 CAS Number: 714971-09-2 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...