R788 disodium salt ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
Sodium (6-((5-fluoro-2-((3,4,5-trimethoxyphenyl)amino)pyrimidin-4-yl)amino)-2,2-dimethyl-3-oxo-2,3-dihydro-4H-pyrido[3,2-b][1,4]oxazin-4-yl)methyl phosphate
, Fostamatinib
SYN-1122-M005
SYN-1122-M001
SYN-1122-M010
SYN-1122-M050
102992-190EA
156.46
USD
102992-190
102992-194
102992-192
102992-196
R788 disodium salt ≥95%
R788 disodium salt
Tamatinib effectively inhibits BCR signaling in vivo, resulting in reduced proliferation and survival of the malignant B cells and significantly prolonged survival of the treated animals.
Formula:
C₂₃H₂₄FN₆O₉P·2Na MW: 624.5 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD15146370 CAS Number: 1025687-58-4 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...