PIK-93 ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
N-(5-(4-Chloro-3-(N-(2-hydroxyethyl)sulphamoyl)phenyl)-4-methylthiazol-2-yl)acetamide
, N-(5-(4-Chloro-3-(N-(2-hydroxyethyl)sulfamoyl)phenyl)-4-methylthiazol-2-yl)acetamide
SYN-1066-M005
SYN-1066-M001
SYN-1066-M050
SYN-1066-M010
102991-768EA
156.46
USD
102991-768
102991-774
102991-770
102991-772
PIK-93 ≥95%
PIK-93
PIK-93 selectively inhibits the type III PI 4-kinase beta enzyme (IC(50) of 19nM). It has also been shown to have activity on PI3Kalpha with an IC(50) of 39nM.
Formula:
C₁₄H₁₆ClN₃O₄S₂ MW: 389.9 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12922510 CAS Number: 593960-11-3 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...