PI103 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 3-(4-Morpholinopyrido[3',2':4,5]furo[3,2-d]pyrimidin-2-yl)phenol

SYN-1065-M005 SYN-1065-M050 SYN-1065-M001 SYN-1065-M010
102991-766EA 672.92 USD
102991-766 102991-762 102991-764 102991-760
PI103 ≥95%
PI103

PI-103 is an isoform-selective class I PI3K and mTOR inhibitor. It has activity on p110alpha-delta with IC(50) values of 2nM, 3nM, 3nM, 15nM respectively. It is less potent against mTOR and DNA-PK with IC(50) values of 30nM and 23nM respectively.

Formula: C₁₉H₁₆N₄O₃
MW: 348.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11983145
CAS Number: 371935-74-9

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR