PD-173955-Analog1 ≥95%
Supplier: ADIPOGEN CORP MS
SYN-1062-M010
SYN-1062-M001
SYN-1062-M005
102991-738EA
224.31
USD
102991-738
102991-740
102991-742
PD-173955-Analog1 ≥95%
PD-173955-Analog1
PD173955-Analog1 is a potent c-Src inhibitor with an IC(50) value of 9nM. It is also a weaker inhibitor of PDGFR, FGFR and EGFR with IC(50) values of 130nM, 110nM and 190nM respectively.
Formula:
C₂₁H₁₄Cl₂N₄O₃ Storage Temperature: Freezer |
CAS Number:
185039-99-0 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO or ethanol. Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...