Masatinib ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
AB1010
SYN-1053-M010
SYN-1053-M050
SYN-1053-M005
SYN-1053-M001
102991-668EA
523.38
USD
102991-668
102991-664
102991-666
102991-662
Masatinib ≥95%
Masatinib
Masitinib is a potent, orally available protein tyrosine kinase inhibitor targeting c-Kit, PDGFR and FGFR3. It demonstrates activity for Kit and PDGFRalpha/beta with IC(50) values of 200nM, 540nM and 800nM respectively. It shows weak inhibition of ABL and c-Fms.
Formula:
C₂₈H₃₀N₆OS MW: 498.7 g/mol Storage Temperature: Freezer |
CAS Number:
790299-79-5 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...