KU-0063794 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
(5-(2-(cis-2,6-Dimethylmorpholino)-4-morpholinopyrido[2,3-d]pyrimidin-7-yl)-2-methoxyphenyl)methanol
SYN-1050-M001
SYN-1050-M050
SYN-1050-M005
SYN-1050-M010
102991-638EA
105.23
USD
102991-638
102991-642
102991-644
102991-640
KU-0063794 ≥95%
KU-0063794
KU0063794 is a highly specific inhibitor of the mammalian target of rapamycin (mTOR). It inhibits both mTORC1 and mTORC2 with an IC(50) of approximately 10nM.
Formula:
C₂₅H₃₁N₅O₄ MW: 465.6 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD22666594 CAS Number: 938440-64-3 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...