GW-441756 ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
3-((1-Methyl-1H-indol-3-yl)methylene)-1,3-dihydro-2H-pyrrolo[3,2-b]pyridin-2-one
SYN-1044-M005
SYN-1044-M050
SYN-1044-M001
SYN-1044-M010
102991-590EA
90
USD
102991-590
102991-592
102991-594
102991-596
GW-441756 ≥95%
GW-441756
GW441756 is an orally active, potent and highly selective TrkA tyrosine kinase inhibitor (IC(50) of 2nM). It specifically blocks TrkA-induced cell death in a dose-dependent manner.
Formula:
C₁₇H₁₃N₃O MW: 275.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD17010281 CAS Number: 504433-23-2 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...