BX-912 ≥95%
Supplier: ADIPOGEN CORP MS
SYN-1027-M005
SYN-1027-M010
SYN-1027-M001
SYN-1027-M050
102991-460EA
956.77
USD
102991-460
102991-458
102991-454
102991-456
BX-912 ≥95%
BX-912
BX912 is a PDK1 (3-Phosphoinositide-dependent Kinase-1) inhibitor, with an IC(50) of 26nM.
Formula:
C₂₀H₂₃BrN₈O MW: 471,4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD10565925 CAS Number: 702674-56-4 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...