Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bosutinib ≥95%

Warning
Synonyms: SKI-606

SYN-1025-M010 SYN-1025-M005 SYN-1025-M050 SYN-1025-M001
102991-442EA 105.23 USD InStock
102991-442 102991-440 102991-444 102991-438
Bosutinib ≥95%
Bosutinib
Bosutinib is a novel, dual Src/Abl inhibitor with IC(50) values of 1.2nM and 1nM, respectively.


Formula: C₂₆H₂₉Cl₂N₅O₃
MW: 530.5 g/mol
Storage Temperature: Freezer
CAS Number: 380843-75-4
Order Now
Specification Test Results
Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty