BEZ235 ≥95%
Synonyms:
NVP-BEZ235
NVP-BEZ235 is an imidazo[4,5-c]quinoline derivative that inhibits PI3K and mTOR kinase activity by binding to the ATP-binding cleft of these enzymes. In cellular settings using human tumor cell lines, this molecule is able to effectively and specifically block the dysfunctional activation of the PI3K pathway, inducing G1 arrest at IC(50) values 3-10nM in biological cell growth and viability assays. In addition NVP-BEZ235 shows effective IC(50) values in in vitro tests against mTor and PI3K family members of 10nM.
Formula:
C₃₀H₂₃N₅O MW: 469.6 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD10565944 CAS Number: 915019-65-7 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...