Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IAXO-101 (CD14/TLR4 Antagonist) (synthetic) ≥98% (by TLC)

Supplier: ADIPOGEN CORP MS
Synonyms: Methyl 6-deoxy-6-N-dimethyl-N-cyclopentylammonium-2,3-di-O-tetradecyl-alpha-D-glucopyranoside iodide

IAX600001M001 IAX600001M005
CD14/TLR4 antagonist. Inhibitor of sterile inflammation. Synthetic TLR4/CD14 ligand with TLR4 modulating activities in vitro, and conferring protection against TLR4/CD14-mediated tissue damage and inflammation in vivo. Useful to explore CD14- dependent and TLR4-independent pathways and TLR4 activation by endogenous ligands (e.g. hyaluronic acid oligosaccharides, oxLDL, HMGB1) in sterile inflammation. Shown to inhibit neuropathic pain, secondary necrosis of acute drug-induced liver failure and vascular inflammation, and abdominal aortic aneurysm by blocking non-hematopoietic TLR4 signaling. Useful tool, where inhibition of sterile (auto-) inflammation is desired, without compromising TLR4’s key role in the defense of pathogens.


Formula: C₄₂H₈₄INO₅
MW: 810,02g/mol (iodide salt) g/mol
Storage Temperature: Refrigerator
CAS Number: 1202388-64-4
Specification Test Results
Source/Host Synthetic.
Purity >98% (TLC)
Appearance/Color White solid.
Solubility Soluble in Methanol, DMSO and Ethanol 1:1 (vol:vol): >10mM.


 

ORDER

Expand All / Collapse All
Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
1 mg Plastic vial IAX600001M001 102991-276 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

5 mg Plastic vial IAX600001M005 102991-278 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty