ZnAF-2F DA ≥95% (by HPCE)

Supplier: ADIPOGEN CORP MS

Synonyms: 6-{2-[Bis(2-pyridylmethyl)amino]ethylamino}-2′,7′-difluorofluorescein diacetate

CDX-Z0012-M001 CDX-Z0012-M005
102991-250EA 193.85 USD
102991-250 102991-252
ZnAF-2F DA ≥95% (by HPCE)
ZnAF-2F DA

For application to living cells, the diacetyl derivative of ZnAF-2F, ZnAF-2F DA, was synthesized. ZnAF-2F DA can permeate through the cell membrane, and is hydrolyzed by esterase in the cytosol to yield ZnAF-2F, which is retained in the cells. Using ZnAF-2F DA we could measure the changes of intracellular Zn2+ in cultured cells and hippocampal slices. Spectral data: lambdaex 492nm; lambdaem 515nm in PBS.

Formula: C₃₈H₃₀F₂N₄O₇
MW: 692.66 g/mol
Storage Temperature: Freezer
CAS Number: 443302-10-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR