ZnAF-2 DA ≥90% (by HPCE)
Supplier: ADIPOGEN CORP MS
Warning
|
Synonyms:
6-{2-[Bis(2-pyridylmethyl)amino]ethylamino}fluorescein diacetate
CDX-Z0008-M005
CDX-Z0008-M001
102991-234EA
193.85
USD
102991-234
102991-236
ZnAF-2 DA ≥90% (by HPCE)
ZnAF-2 DA
Suitable for the fluorimetric detection of Zn2+ (after esterase hydrolysis). Spectral data: lambdaex 492nm; lambdaem 515nm in PBS.
Formula:
C₃₈H₃₂N₄O₇ MW: 656.68 g/mol Storage Temperature: Freezer |
CAS Number:
357339-96-9 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...