Trimethoprim sulfate ≥99% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
2,4-Pyrimidinediamine, 5-((3,4-trimethoxyphenyl)methyl)-, sulfate (2:1)
CDX-T0146-M100
CDX-T0146-M025
102991-142EA
157.85
USD
102991-142
102991-144
Trimethoprim sulfate ≥99% (by HPLC)
Trimethoprim sulfate
Antibacterial compound. Pyrimidine inhibitor of dihydrofolate reductase.
Formula:
C₂₈H₃₈N₈O₁₀S MW: 678.24 g/mol Storage Temperature: Freezer |
CAS Number:
56585-33-2 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in water. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...