O-Desmethyltramadol hydrochloride ≥98% (by HPLC)
Synonyms:
rel-3-[(1R,2R)-2-[(dimethylamino)methyl]-1-hydroxycyclohexyl]-phenol, monohydrochloride
, O-DSMT
, ODT
CDX-D0219-M025
CDX-D0219-M100
102989-676EA
94
USD
102989-676
102989-678
O-Desmethyltramadol hydrochloride ≥98% (by HPLC)
O-Desmethyltramadol hydrochloride
Analgesic compound. Main active metabolite of tramadol. More potent as a µ: opioid agonist compared to tramadol. High-affinity ligand of the delta- and kappa-opioid receptors. Inactive as serotonin reuptake inhibitor. Compound can be used as analytical reference material.
Formula:
C₁₅H₂₃NO₂·HCl MW: 285.8 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD08063708 CAS Number: 185453-02-5 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in water or methanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...