DAR-1T ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Diaminorhodamine-1T

CDX-D0213-M001 CDX-D0213-M005
102989-672EA 333.69 USD
102989-672 102989-674
DAR-1T ≥95% (by HPLC)
DAR-1T

Reference compound for the fluorescent (nitric oxide) NO probe DAR-1.

Formula: C₂₈H₂₉N₅O₃
MW: 483.57 g/mol
Storage Temperature: Freezer
CAS Number: 261351-46-6

Order Now

Specification Test Results

Source/Host Synthetic
Purity 95% (HPLC)
Appearance/Color Dark violet to black powder.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR