6-Amino TAMRA

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 6-Aminotetramethylrhodamine

CDX-A0157-M010
102989-210EA 276.92 USD
102989-210
6-Amino TAMRA
6-Amino TAMRA

A synthetic intermediate.

Formula: C₂₄H₂₃N₃O₃
MW: 401.46 g/mol
Storage Temperature: Refrigerator
CAS Number: 159435-10-6

Order Now

Specification Test Results

Source/Host Synthetic.
Appearance/Color Solid.
Solubility Soluble in DMSO or methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR