Piperafizine B ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 3,6-Dibenzylidene-2,5-piperazinedione

BVT-0446-C500 BVT-0446-M001
102989-044EA 152.31 USD
102989-044 102989-046
Piperafizine B ≥98% (by HPLC)
Piperafizine B

Diketopiperazine derivative. Analog of Piperafizine A. Potentiator of vincristine cytotoxicity.

Formula: C₁₈H₁₄N₂O₂
MW: 290.3 g/mol
Storage Temperature: Refrigerator
CAS Number: 74720-33-5

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces thioluteus.
Purity 98% (HPLC)
Appearance/Color Light yellow solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR