ADIPAG-CS1-0102
AGCS10102M001 AGCS10102M010
102987-450EA 843.23 USD
102987-450 102987-448
ADIPAG-CS1-0102
Potent, reversible and cell-permeable inhibitor of the 19S deubiquitinases (DUBs) ubiquitin-specific-processing protease 14 (USP14) and ubiquitin carboxyl-terminal hydrolase isozyme L5 (UCHL5). Inhibitor of the 20S proteasome. Anti-tumor compound.