COX-2 Inhibitor 8c ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Compound 8c

AG-CR1-3606-M001
102988-742EA 221.54 USD
102988-742
COX-2 Inhibitor 8c ≥98% (by NMR)
COX-2 Inhibitor 8c

Potent cyclooxygenase-2 (COX-2) inhibitor. Inhibits inflammatory prostaglandin E2 (PGE2) production.

Formula: C₁₂H₁₁NO₂S₂
MW: 265.4 g/mol
Storage Temperature: Freezer
CAS Number: 1259295-44-7

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White solid.
Solubility Soluble in DMSO or methanol. Insoluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR