Malformin A1 ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Peptide antibiotic
Source/Host: Synthetic. Originally isolated from Aspergillus niger.
Purity: >95% (HPLC)
Appearance/Color: White solid.
Solubility: Soluble in DMSO.
Antibcaterial. Plant growth stimulator. Induces root curvature and malformation in plants. Mycotoxin. Prevents interleukin-1 (IL-1) induced endothelial changes by inhibition of protein synthesis. Inhibitor of interleukin-1 beta (IL1 beta) binding to its receptor. Enhancer of cellular fibrinolytic activity. Disrupts the cell cycle at the G2 checkpoint of cancer cells, leading to sensitization of the cancer cells to anti-cancer reagents. Anticancer compound. Cytotoxic against several cancer cell lines. Antimalarial and antitrypanosomal. Inhibitor of BRAF-mutated melanoma cell lines.
Storage Temperature:
Freezer |
CAS Number:
3022-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...