Martinomycin ≥95% (by TLC)

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0159-M005 AG-CN2-0159-M001
102987-414EA 256.15 USD
102987-414 102987-416
Martinomycin ≥95% (by TLC)
Martinomycin

Antibiotic


Source/Host: Isolated from Actinoallamurus yoronensis.
Purity: >95% (TLC)
Appearance/Color: Off-white solid.
Solubility: Soluble in DMSO, ethanol or methanol.


Active against Gram-positive bacteria. Active against Spodoptera eridania.

Storage Temperature: Freezer
CAS Number: 160791-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR