Aspergillimide ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0145-M005 AG-CN2-0145-M001
102988-068EA 830.77 USD
102988-068 102988-066
Aspergillimide ≥95% (by HPLC)
Aspergillimide

Antibiotic


Source/Host: Isolated from Aspergillus japonicus Fg-551.
Purity: >95% (HPLC)
Appearance/Color: White solid.
Solubility: Soluble in methanol, ethyl acetate, acetone or chloroform. Insoluble in water or hexane.


Anthelmintic agent. Paralytic against Bombyx mori.

Storage Temperature: Freezer
CAS Number: 195966-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR