Cyclopenol ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0136-M001
102988-036EA 249.23 USD
102988-036
Cyclopenol ≥95% (by HPLC)
Cyclopenol

Antimicrobial. Phytotoxic.

Formula: C₁₇H₁₄N₂O₄
Storage Temperature: Freezer
CAS Number: 20007-85-6

Order Now

Specification Test Results

Source/Host Isolated from Penicillium sp.
Purity 95% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in DMSO, ethanol or methanol. Insoluble in hexane

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR