Zelkovamycin ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-CN2-0128-M001
AG-CN2-0128-C250
102988-006EA
96.92
USD
102988-006
102988-008
Zelkovamycin ≥95% (by HPLC)
Zelkovamycin
Cyclic peptide antibiotic
Source/Host: Isolated from Streptomyces sp.
Purity: >95% (HPLC)
Appearance/Color: White solid.
Solubility: Soluble in DMSO, ethanol or methanol.
Antibacterial.
Storage Temperature:
Freezer |
CAS Number:
221197-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...