Dihydrochlamydocin ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-CN2-0115-C250
AG-CN2-0115-M001
102987-402EA
96.92
USD
102987-402
102987-404
Dihydrochlamydocin ≥95% (by HPLC)
Dihydrochlamydocin
Phytotoxin. Derivative of HDAC inhibitor chlamydocin Similar chemical structure to the TAN-1746 compounds (potent HDAC inhibitors).
Formula:
C₂₈H₄₀N₄O₆ Storage Temperature: Freezer |
CAS Number:
157618-75-2 |
Specification Test Results
Source/Host | Isolated from Verticillium sp. |
Purity |
|
Appearance/Color | Off-white solid. |
Solubility | Soluble in DMSO, ethanol or methanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...